Learn More
TOKU-E Bleomycin ReadyMade™ Solution | Bleomycin Sulfate | CAS 9041-93-4 | 10 mg/ML in water | sterile-filtered | 10 x 1 ML
Catalog No. NC2068739
Encompass_Preferred
Bleomycin ReadyMade™ Solution | Bleomycin Sulfate | CAS 9041-93-4 | 10 mg/ML in water | sterile-filtered | 10 x 1 ML
Catalog No. NC2068739 Supplier TOKU-E Supplier No. B09610X1ML
Bleomycin ReadyMade™ Solution | Bleomycin Sulfate | CAS 9041-93-4 | 10 mg/ML in water | sterile-filtered | 10 x 1 ML