Learn More
Lkt Laboratories CAS 9041-93-4, ≥90%, Mixture of glycopeptide bleomycin sulfate salts that induces DNA strand breaks and is used to treat various cancers.
SDP
Catalog No. NC2539802
Encompass_Preferred
This product is a mixture of bleomycin sulfate salts; bleomycin A2 is pictured. Bleomycin is an anticancer chemotherapeutic that binds DNA and induces DNA cleavage and strand breaks; it is highly toxic. Bleomycin A5 is clinically used to treat lymphomas, squamous cell carcinomas, and testicular cancer. Additionally, this compound induces apoptosis, alters cell cycle regulation, inhibits proliferation, and decreases tumor size in cellular and animal models of hemangioma. Bleomycin A2 inhibits viral reproduction of human papilloma virus. Bleomycin A2 also cleaves DNA at 5’-GT sequences (including in telomeres) and induces tRNA cleavage. Bleomycin may also inhibit thioredoxin reductase.
Catalog No. NC2539802 Supplier Lkt Laboratories Supplier No. B4518
This product is a mixture of bleomycin sulfate salts; bleomycin A2 is pictured. Bleomycin is an anticancer chemotherapeutic that binds DNA and induces DNA cleavage and strand breaks; it is highly toxic. Bleomycin A5 is clinically used to treat lymphomas, squamous cell carcinomas, and testicular cancer. Additionally, this compound induces apoptosis, alters cell cycle regulation, inhibits proliferation, and decreases tumor size in cellular and animal models of hemangioma. Bleomycin A2 inhibits viral reproduction of human papilloma virus. Bleomycin A2 also cleaves DNA at 5’-GT sequences (including in telomeres) and induces tRNA cleavage. Bleomycin may also inhibit thioredoxin reductase.