Learn More
Selleck Chemical LLC Bleomycin sulfate S1214-1g
SDP
Catalog No. 503111148
Encompass_Preferred
Bleomycin sulfate is a glycopeptide antibiotic and a DNA synthesis inhibitor as well as an anticancer agent for squamous cell carcinomas (SCC) with an IC50 of 4 nM in UT-SCC-19A cells Bleomycin sulfate is a DNA synthesis inhibitor Bleomycin (NSC125066) sulfate can be used to induce animal models of pulmonary fibrosis
Catalog No. 50-311-1148 Supplier Selleck Chemical LLC Supplier No. S12141G
Bleomycin sulfate is a glycopeptide antibiotic and a DNA synthesis inhibitor as well as an anticancer agent for squamous cell carcinomas (SCC) with an IC50 of 4 nM in UT-SCC-19A cells Bleomycin sulfate is a DNA synthesis inhibitor Bleomycin (NSC125066) sulfate can be used to induce animal models of pulmonary fibrosis