Learn More
Selleck Chemical LLC Bleomycin sulfate S1214-200mg
SDP
Catalog No. 503111419
Encompass_Preferred
Bleomycin sulfate is a glycopeptide antibiotic and a DNA synthesis inhibitor as well as an anticancer agent for squamous cell carcinomas (SCC) with an IC50 of 4 nM in UT-SCC-19A cells Bleomycin sulfate is a DNA synthesis inhibitor Bleomycin (NSC125066) sulfate can be used to induce animal models of pulmonary fibrosis
Catalog No. 50-311-1419 Supplier Selleck Chemical LLC Supplier No. S1214200MG
Bleomycin sulfate is a glycopeptide antibiotic and a DNA synthesis inhibitor as well as an anticancer agent for squamous cell carcinomas (SCC) with an IC50 of 4 nM in UT-SCC-19A cells Bleomycin sulfate is a DNA synthesis inhibitor Bleomycin (NSC125066) sulfate can be used to induce animal models of pulmonary fibrosis