Learn More
Research Products International Corp Bleomycin Sulfate, 5 Milligrams
SDP
Catalog No. 50488538
Encompass_Preferred
Bleomycin sulfate is an antibiotic derived from Streptomyces verticillus. It has found application in the field of cancer research and treatment. This antibiotic is a glycopeptide with DNA-cleaving properties. It is known for its ability to induce DNA strand breaks, making it effective in inhibiting the replication of cancer cells. 
In research settings, bleomycin sulfate is utilized as a tool to study DNA damage and repair mechanisms. It is commonly employed in cell biology experiments to induce controlled DNA damage for the investigation of cellular responses and signaling pathways associated with DNA repair.
Catalog No. 50-488-538 Supplier Research Products International Corp Supplier No. B400600.005
Bleomycin sulfate is an antibiotic derived from Streptomyces verticillus. It has found application in the field of cancer research and treatment. This antibiotic is a glycopeptide with DNA-cleaving properties. It is known for its ability to induce DNA strand breaks, making it effective in inhibiting the replication of cancer cells. 
In research settings, bleomycin sulfate is utilized as a tool to study DNA damage and repair mechanisms. It is commonly employed in cell biology experiments to induce controlled DNA damage for the investigation of cellular responses and signaling pathways associated with DNA repair.