Learn More
Enzo Life Sciences Bleomycin sulfate, (50mg), CAS Number: 9041-93-4
SDP
Catalog No. NC9949864
Encompass_Preferred
Antitumor antibiotic. Anti-tumor agent. Induces apoptosis via DNA damage. Radiomimetic DNA-cleaving agent that produces double and single DNA strand breaks through an oxygen-radical-dependent mechanism. Inhibits intracellular DNA replication. DNA alkylating agent. Alternative Name: Blenoxane. Formula: C55H85N17O25S4. MW: 1512.6. CAS Number: 9041-93-4. MI: 14: 1318. Purity: ≥98% (HPLC). Appearance: White crystalline solid. Solubility: Soluble in water (20mg/ml) or methanol. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC9949864 Supplier Enzo Life Sciences Supplier No. BMLAP3020050
Antitumor antibiotic. Anti-tumor agent. Induces apoptosis via DNA damage. Radiomimetic DNA-cleaving agent that produces double and single DNA strand breaks through an oxygen-radical-dependent mechanism. Inhibits intracellular DNA replication. DNA alkylating agent. Alternative Name: Blenoxane. Formula: C55H85N17O25S4. MW: 1512.6. CAS Number: 9041-93-4. MI: 14: 1318. Purity: ≥98% (HPLC). Appearance: White crystalline solid. Solubility: Soluble in water (20mg/ml) or methanol. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C