Learn More
Medchemexpress LLC Bleomycin (sulfate) | 9041-93-4 | 99.49% | 1523 (average) | 100 MG
SDP
Catalog No. 5000387338
Encompass_Preferred
Bleomycin sulfate is a DNA synthesis inhibitor and an antitumor antibiotic. It acts as a DNA damaging agent, causing single and double strand breaks and single apurinic/apyrimidinic sites in DNA. This radiomimetic compound is used to induce pulmonary fibrosis models and is rapidly absorbed with high bioavailability.
  • DNA synthesis inhibitor
  • Antitumor antibiotic
  • Radiomimetic compound, resembling the genetic effect of ionizing radiation
  • Causes single and double strand breaks and single apurinic/apyrimidinic sites in DNA
  • Can be used to induce pulmonary fibrosis models
  • Rapidly absorbed after intramuscular, subcutaneous, intraperitoneal, or intrapleural administration
  • Reaches peak plasma concentrations in approximately 60 minutes
  • Less than 1% binds to plasma proteins
  • High bioavailability
  • Mean plasma clearance rate of about 70 mL/min/m2
Catalog No. 50-003-87338 Supplier Medchemexpress LLC Supplier No. HY17565100MG
Bleomycin sulfate is a DNA synthesis inhibitor and an antitumor antibiotic. It acts as a DNA damaging agent, causing single and double strand breaks and single apurinic/apyrimidinic sites in DNA. This radiomimetic compound is used to induce pulmonary fibrosis models and is rapidly absorbed with high bioavailability.
  • DNA synthesis inhibitor
  • Antitumor antibiotic
  • Radiomimetic compound, resembling the genetic effect of ionizing radiation
  • Causes single and double strand breaks and single apurinic/apyrimidinic sites in DNA
  • Can be used to induce pulmonary fibrosis models
  • Rapidly absorbed after intramuscular, subcutaneous, intraperitoneal, or intrapleural administration
  • Reaches peak plasma concentrations in approximately 60 minutes
  • Less than 1% binds to plasma proteins
  • High bioavailability
  • Mean plasma clearance rate of about 70 mL/min/m2