Learn More
Medchemexpress LLC Bleomycin (sulfate) | 9041-93-4 | 99.9% | 1523 | 500 MG
SDP
Catalog No. 5000387463
Encompass_Preferred
Bleomycin sulfate is a potent DNA synthesis inhibitor and antitumor antibiotic, recognized for its role as a DNA damaging agent. It is widely used in research for its biological activity as a micronucleus inducer in human lymphocytes and its ability to induce pulmonary fibrosis models.
  • DNA synthesis inhibitor and antitumor antibiotic
  • Micronucleus inducer in human lymphocytes
  • Induces single and double-strand breaks in DNA
  • Radiomimetic compound, mimicking genetic effect of ionizing radiation
  • Used to induce pulmonary fibrosis models in animal studies
  • Rapidly absorbed with high bioavailability
  • High plasma elimination and urinary excretion rates
Catalog No. 50-003-87463 Supplier Medchemexpress LLC Supplier No. HY17565500MG
Bleomycin sulfate is a potent DNA synthesis inhibitor and antitumor antibiotic, recognized for its role as a DNA damaging agent. It is widely used in research for its biological activity as a micronucleus inducer in human lymphocytes and its ability to induce pulmonary fibrosis models.
  • DNA synthesis inhibitor and antitumor antibiotic
  • Micronucleus inducer in human lymphocytes
  • Induces single and double-strand breaks in DNA
  • Radiomimetic compound, mimicking genetic effect of ionizing radiation
  • Used to induce pulmonary fibrosis models in animal studies
  • Rapidly absorbed with high bioavailability
  • High plasma elimination and urinary excretion rates