Learn More
Medchemexpress LLC Bleomycin (sulfate) | 9041-93-4 | 99.9% | 1523 (average) | 5 MG
SDP
Catalog No. 5000387464
Encompass_Preferred
Bleomycin sulfate is a potent DNA synthesis inhibitor and an antitumor antibiotic. This compound is known for its DNA damaging properties and its ability to closely mimic the genetic effects of ionizing radiation, making it a valuable tool in research.
  • Mechanism of action: DNA synthesis inhibitor and antitumor antibiotic.
  • Induces micronuclei in human lymphocytes.
  • Causes single and double-strand DNA breaks.
  • Considered a radiomimetic compound.
  • Can be used to induce pulmonary fibrosis models in vivo.
  • Rapidly absorbed with high bioavailability after various administration routes.
  • High plasma elimination and urinary excretion rates.
Catalog No. 50-003-87464 Supplier Medchemexpress LLC Supplier No. HY175655MG
Bleomycin sulfate is a potent DNA synthesis inhibitor and an antitumor antibiotic. This compound is known for its DNA damaging properties and its ability to closely mimic the genetic effects of ionizing radiation, making it a valuable tool in research.
  • Mechanism of action: DNA synthesis inhibitor and antitumor antibiotic.
  • Induces micronuclei in human lymphocytes.
  • Causes single and double-strand DNA breaks.
  • Considered a radiomimetic compound.
  • Can be used to induce pulmonary fibrosis models in vivo.
  • Rapidly absorbed with high bioavailability after various administration routes.
  • High plasma elimination and urinary excretion rates.