Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bleomycin Sulfate, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139824
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139824 50 mg
S000039 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139824 Supplier G-Biosciences Supplier No. RC901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bleomycin Sulfate is an anti-cancer agent isolated from Streptomyces verticillus. Bleomycin is a mixture of bleomycin A2 and bleomycin B2, in an approximately 2:1 ratio, respectively.

Bleomycin is used against Hodgkin's lymphoma. The enzymes react with oxygen and produce free radicals, ultimately causing single-stranded breaks in deoxyribose sugars of DNA.

Chemical Identifiers

Molecular Weight (g/mol) 1512.62 g/mol

Specifications

Quantity 50 mg
Percent Purity 98%
Grade USP
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.