Learn More
Cayman Chemical Bleomycin (sulfate), Purity: ≥95% (mixture of A2 and B2), CAS: 9041-93-4, 50 mg
SDP
Catalog No. 502954353
Encompass_Preferred
A glycopeptide antitumor antibiotic that produces single and double-strand DNA breaks in tumor cells to interrupt their cell cycle; has been used for the treatment of Hodgkin’s lymphoma in combination with doxorubicin, squamous cell carcinomas, testicular cancer, as well as in animal models of pulmonary fibrosis
Catalog No. 50-295-4353 Supplier Cayman Chemical Supplier No. 1387750 MG
A glycopeptide antitumor antibiotic that produces single and double-strand DNA breaks in tumor cells to interrupt their cell cycle; has been used for the treatment of Hodgkin’s lymphoma in combination with doxorubicin, squamous cell carcinomas, testicular cancer, as well as in animal models of pulmonary fibrosis