Learn More
Medchemexpress LLC Bleximenib oxalate | 2866179-95-3 | 98.0% | 689.82 | 25 MG
SDP
Catalog No. 5000363380
Encompass_Preferred
Bleximenib oxalate is an orally active and selective menin-KMT2A inhibitor. It inhibits proliferation, induces apoptosis and differentiation of tumor cells, and is used in the research of tumors such as leukemia.
  • Reduces gene expression of MEIS1, PBX3, FLT3, JMJD1C
  • Inhibits proliferation of leukemia cells
  • Induces apoptosis and differentiation
  • Exhibits anti-tumor activity in leukemia mouse models
  • Increases lifespan in mouse models
Catalog No. 50-003-63380 Supplier Medchemexpress LLC Supplier No. 25MGHY148669A
Bleximenib oxalate is an orally active and selective menin-KMT2A inhibitor. It inhibits proliferation, induces apoptosis and differentiation of tumor cells, and is used in the research of tumors such as leukemia.
  • Reduces gene expression of MEIS1, PBX3, FLT3, JMJD1C
  • Inhibits proliferation of leukemia cells
  • Induces apoptosis and differentiation
  • Exhibits anti-tumor activity in leukemia mouse models
  • Increases lifespan in mouse models