Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bleximenib oxalate | 2866179-95-3 | 98.0% | 689.82 | C34H52FN7O7 | 10 MG
SDP

Catalog No. 5000285608
Encompass_Preferred

Bleximenib oxalate is the oxalate salt of bleximenib, an orally active and selective menin-KMT2A inhibitor used for preclinical oncology research. The compound is supplied as a light yellow solid with manufacturer-reported purity, solubility, and recommended storage conditions to support experimental handling and formulation.

  • Purity 98.0%.
  • Light yellow solid physical form.
  • Selective menin-KMT2A inhibitor for preclinical studies.
  • Soluble in DMSO (100 mg/mL) with published in vivo formulation protocols.
  • Recommended storage for powder and solutions to maintain stability.
  • Available in small research pack sizes for dosing flexibility.

Catalog No. 50-002-85608 Supplier Medchemexpress LLC Supplier No. HY148669A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bleximenib oxalate is the oxalate salt of bleximenib, an orally active and selective menin-KMT2A inhibitor used for preclinical oncology research. The compound is supplied as a light yellow solid with manufacturer-reported purity, solubility, and recommended storage conditions to support experimental handling and formulation.

  • Purity 98.0%.
  • Light yellow solid physical form.
  • Selective menin-KMT2A inhibitor for preclinical studies.
  • Soluble in DMSO (100 mg/mL) with published in vivo formulation protocols.
  • Recommended storage for powder and solutions to maintain stability.
  • Available in small research pack sizes for dosing flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.