Learn More
Azzota Corp BLF200 UC 10L 35kHz 200W drain
Catalog No. 5000887242
Encompass_Preferred
Model BLF200 Low-frequency Fixed Power Ultrasonic Cleaners - Benchtop 35 KHZ 10L 200W 30x24x15cm 1 - 99 Min With Drain
Catalog No. 50-008-87242 Supplier Azzota Corp Supplier No. BLF200
Model BLF200 Low-frequency Fixed Power Ultrasonic Cleaners - Benchtop 35 KHZ 10L 200W 30x24x15cm 1 - 99 Min With Drain