Learn More
Medchemexpress LLC BLI-489 hydrate 1mg | 2580120-08-5 | 1MG
SDP
Catalog No. 5000241730
Encompass_Preferred
BLI-489 hydrate a penem -lactamase inhibitor is active against class A and class C as well as some class D -lactamases The combination of Piperacillin and BLI-489 hydrate is efficacious against murine infections caused by class A (including extended-spectrum -lactamases) class C (AmpC) and class D -lactamase-expressing pathogens[1 [2
Catalog No. 50-002-41730 Supplier Medchemexpress LLC Supplier No. HY108062A1MG
BLI-489 hydrate a penem -lactamase inhibitor is active against class A and class C as well as some class D -lactamases The combination of Piperacillin and BLI-489 hydrate is efficacious against murine infections caused by class A (including extended-spectrum -lactamases) class C (AmpC) and class D -lactamase-expressing pathogens[1 [2