Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BLI-489 hydrate | 623564-40-9 | MFCD22374169 | 95.8% | 327.29 | C13H10N3NaO4S | 25 MG
SDP

Catalog No. 5000241717
Encompass_Preferred

BLI-489 is a penem-class beta-lactamase inhibitor supplied for research use. It is provided as a sodium salt hydrate in a light yellow to yellow solid form with reported purity of 95.77%, and is offered in both solid packs and as a 10 mM solution for assay applications.

  • Beta-lactamase inhibitor active against class A, C, and D enzymes
  • Supplied as a light yellow to yellow solid suitable for sealed storage
  • Reported purity 95.8%
  • Molecular weight 327.29 g/mol (anhydrous basis)
  • Available in small research pack sizes and as a 10 mM solution in DMSO
  • Storage: solids at 4°C sealed; solutions at -20°C to -80°C depending on duration

Catalog No. 50-002-41717 Supplier Medchemexpress LLC Supplier No. HY10806225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

BLI-489 is a penem-class beta-lactamase inhibitor supplied for research use. It is provided as a sodium salt hydrate in a light yellow to yellow solid form with reported purity of 95.77%, and is offered in both solid packs and as a 10 mM solution for assay applications.

  • Beta-lactamase inhibitor active against class A, C, and D enzymes
  • Supplied as a light yellow to yellow solid suitable for sealed storage
  • Reported purity 95.8%
  • Molecular weight 327.29 g/mol (anhydrous basis)
  • Available in small research pack sizes and as a 10 mM solution in DMSO
  • Storage: solids at 4°C sealed; solutions at -20°C to -80°C depending on duration

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.