Learn More
MSC Blickle Caster, 4 in, Thermoplastic Polyurethane, Total-Lock Brake
Catalog No. 5000534944
Encompass
This Blickle swivel caster features a 4 inch diameter wheel made of thermoplastic polyurethane, designed for smooth running performance and noise absorption. It is equipped with a total-lock brake, providing secure positioning for various applications.
  • Smooth running performance
  • Noise absorbing design
  • Elastic solid rubber damping ring
  • Very high abrasion resistance
  • Suitable for medium-duty and heavy-duty applications
  • Sealed ball bearing for durability
  • Zinc finish
Catalog No. 50-005-34944 Supplier MSC Supplier No. 59187062
This Blickle swivel caster features a 4 inch diameter wheel made of thermoplastic polyurethane, designed for smooth running performance and noise absorption. It is equipped with a total-lock brake, providing secure positioning for various applications.
  • Smooth running performance
  • Noise absorbing design
  • Elastic solid rubber damping ring
  • Very high abrasion resistance
  • Suitable for medium-duty and heavy-duty applications
  • Sealed ball bearing for durability
  • Zinc finish