Learn More
Chemglass Life Sciences M12 Blind Stopper, for Bio Reactors
SDP
Catalog No. 501294426
Encompass
M12 Blind Stopper. Stoppers can be used to close off any of the unused openings in the head plate. The M6, M10 and M12 sizes are fitted into the head plate from the inside, leaving a flat surface, while the M18, M27 and G3/4 are threaded into the ports from the outside.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
This item is not returnable. View return policy
Catalog No. 50-129-4426 Supplier Chemglass Life Sciences Supplier No. CLS138024
M12 Blind Stopper. Stoppers can be used to close off any of the unused openings in the head plate. The M6, M10 and M12 sizes are fitted into the head plate from the inside, leaving a flat surface, while the M18, M27 and G3/4 are threaded into the ports from the outside.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.