Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bliretrigine | 1233229-75-8 | 99.4% | C20H24N4O2 | 10MG
SDP

Catalog No. 5000259068
Encompass_Preferred

Bliretrigine is a small-molecule sodium channel blocker used in research with reported analgesic effects. The product is provided as a white to off-white solid with specified solubility in DMSO and defined storage recommendations for both powder and solution forms.

  • Sodium channel blocker with analgesic activity.
  • Molecular formula C20H24N4O2.
  • Molecular weight 352.43.
  • Purity 99.42%.
  • Appearance white to off-white solid.
  • Soluble in DMSO at 50 mg/mL; may require ultrasonic assistance.
  • Store powder at -20°C for up to 3 years or at 4°C for up to 2 years.
  • Store in solution at -80°C for up to 6 months or at -20°C for up to 1 month.

Catalog No. 50-002-59068 Supplier Medchemexpress LLC Supplier No. HY14555810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bliretrigine is a small-molecule sodium channel blocker used in research with reported analgesic effects. The product is provided as a white to off-white solid with specified solubility in DMSO and defined storage recommendations for both powder and solution forms.

  • Sodium channel blocker with analgesic activity.
  • Molecular formula C20H24N4O2.
  • Molecular weight 352.43.
  • Purity 99.42%.
  • Appearance white to off-white solid.
  • Soluble in DMSO at 50 mg/mL; may require ultrasonic assistance.
  • Store powder at -20°C for up to 3 years or at 4°C for up to 2 years.
  • Store in solution at -80°C for up to 6 months or at -20°C for up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.