Learn More
Nuaire Inc BLIZZARD NU-99729VFT -86 CEL
SDP
Catalog No. NC3355228
Encompass
  • Temperature range of -86 °C for critical sample storage.
  • Variable Frequency Drive (VFD) enhances energy efficiency and reduces noise.
  • Intuitive touchscreen control panel for easy operation.
  • Performance tracking for up to ten years.
  • Digital inventory management for sample tracking.
  • Dual-compression technology ensures sample safety.
  • Vacuum-insulated panels (VIP) provide superior insulation.
  • Quadruple-gasketed outer door to maintain temperature control.
  • Environmentally friendly HC refrigerants for green lab initiatives.
  • Net weight of 771 lbs (350 kg) for substantial stability.
  • ENERGY STAR certified in the US and Canada for energy savings.
  • 3-year parts and labor warranty in the US & CA.
Catalog No. NC3355228 Supplier Nuaire Inc Supplier No. NU99729VFT
Add to Cart
Add to Cart
Request a Quote
  • Temperature range of -86 °C for critical sample storage.
  • Variable Frequency Drive (VFD) enhances energy efficiency and reduces noise.
  • Intuitive touchscreen control panel for easy operation.
  • Performance tracking for up to ten years.
  • Digital inventory management for sample tracking.
  • Dual-compression technology ensures sample safety.
  • Vacuum-insulated panels (VIP) provide superior insulation.
  • Quadruple-gasketed outer door to maintain temperature control.
  • Environmentally friendly HC refrigerants for green lab initiatives.
  • Net weight of 771 lbs (350 kg) for substantial stability.
  • ENERGY STAR certified in the US and Canada for energy savings.
  • 3-year parts and labor warranty in the US & CA.