Learn More
Sino Biological Recombinant Human BLK Protein (Full Length, GST Tag), Active
Catalog No. 502569729
Encompass_Preferred
Recombinant full length human BLK was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Catalog No. 50-256-9729 Supplier Sino Biological Supplier No. B0210G10
Recombinant full length human BLK was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.