Learn More
Perkin Elmer US LLC Flared PVC Two-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 12
Catalog No. 509052407
Encompass_Preferred
This flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-905-2407 Supplier Perkin Elmer US LLC Supplier No. N0777043
This flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.