Learn More
Perkin Elmer US LLC Flared PVC MP2 Two-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 12
Catalog No. NC0013512
Encompass_Preferred
MPP PVC Tubing Flared, 0. 76mm, black black, 12/pk. Flared tubing is easier to change and more reliably connected than non-flared small-bore tubing.
Catalog No. NC0013512 Supplier Perkin Elmer US LLC Supplier No. N8145202
MPP PVC Tubing Flared, 0. 76mm, black black, 12/pk. Flared tubing is easier to change and more reliably connected than non-flared small-bore tubing.