Learn More
Genetex Inc GeneTex Blocking Buffer, 5% Milk For WB and ELISA, Reduces Background Interference, 250 mL
SDP
Catalog No. NC2323533
Encompass
GeneTex Blocking Buffer, TBS with 5% Milk is a ready-to-use solution that enhances assay sensitivity by minimizing non-specific background interference in immunological detection methods. It is specifically validated for Western blotting (WB) and ELISA, improving signal-to-noise ratios by binding to sites of nonspecific interaction. This liquid formulation comes in a 250 mL volume, making it ideal for various applications including immunohistochemistry.
Catalog No. NC2323533 Supplier Genetex Inc Supplier No. GTX48883
GeneTex Blocking Buffer, TBS with 5% Milk is a ready-to-use solution that enhances assay sensitivity by minimizing non-specific background interference in immunological detection methods. It is specifically validated for Western blotting (WB) and ELISA, improving signal-to-noise ratios by binding to sites of nonspecific interaction. This liquid formulation comes in a 250 mL volume, making it ideal for various applications including immunohistochemistry.