Learn More
Thomas Scientific 96W Plate, Polypropylene, 370µL/well, Flat Bottom, Chimney Style, Black, 100/case
SDP
Catalog No. 50320828
Encompass_Preferred
Microplates are manufactured from ClearChoice™ virgin polypropylene resin. They are produced with uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics. These plates are biologically inert, exhibit low binding characteristics, offer high temperature tolerance (–196 to 121°C/–321 to 250°F), and are resistant to many standard laboratory chemicals. Black plates are ideal for fluorescence polarization. Microplates feature alphanumerically coded wells with round, conical, or flat bottoms. Chimney well plates feature a chimney-like arrangement in which each well stands on its own to reduce the possibility of cross-contamination, and feature individual rims that rise above the surface of the plate to aid in sealing by cap mat, thermal sealer, or adhesive sealer. Stackable and autoclavable. Plates are supplied without lids. Dimensions: 128Lx86Wx15Hmm.
This item is not returnable. View return policy
Catalog No. 50-320-828 Supplier Thomas Scientific Supplier No. 1184X18
Microplates are manufactured from ClearChoice™ virgin polypropylene resin. They are produced with uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics. These plates are biologically inert, exhibit low binding characteristics, offer high temperature tolerance (–196 to 121°C/–321 to 250°F), and are resistant to many standard laboratory chemicals. Black plates are ideal for fluorescence polarization. Microplates feature alphanumerically coded wells with round, conical, or flat bottoms. Chimney well plates feature a chimney-like arrangement in which each well stands on its own to reduce the possibility of cross-contamination, and feature individual rims that rise above the surface of the plate to aid in sealing by cap mat, thermal sealer, or adhesive sealer. Stackable and autoclavable. Plates are supplied without lids. Dimensions: 128Lx86Wx15Hmm.