Learn More
Bellco Glass, Inc. Blk Phenolic Screw Cap, 13mm Long Skirted - With Liner
SDP
Catalog No. 501945326
Encompass_Preferred
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. THREAD FINISH:GPI 13-415, DIMENSIONS: OD OF CAP; 16.1mm, APPROX, ID OF CAP, 11.6mm, APPROX, LENGTH: 13.7mm, APPROX.
Catalog No. 50-194-5326 Supplier Bellco Glass, Inc. Supplier No. 202900013
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. THREAD FINISH:GPI 13-415, DIMENSIONS: OD OF CAP; 16.1mm, APPROX, ID OF CAP, 11.6mm, APPROX, LENGTH: 13.7mm, APPROX.