Learn More
Bellco Glass, Inc. Blk Phenolic Screw Cap, 15mm Long Skirted - With Liner
SDP
Catalog No. 501945327
Encompass_Preferred
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 15-415, DIMENSIONS: OD OF CAP; 17.7mm, APPROX, ID OF CAP, 13.3mm, APPROX, LENGTH: 15.8mm, APPROX.
Catalog No. 50-194-5327 Supplier Bellco Glass, Inc. Supplier No. 202900016
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 15-415, DIMENSIONS: OD OF CAP; 17.7mm, APPROX, ID OF CAP, 13.3mm, APPROX, LENGTH: 15.8mm, APPROX.