Learn More
Bellco Glass, Inc. Blk Phenolic Screw Cap, 18mm Long Skirted - With Liner
SDP
Catalog No. 501945328
Encompass_Preferred
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 18-415, DIMENSIONS: OD OF CAP: 20.5mm, APPROX, ID OF CAP: 16.1mm, APROX, LENGTH: 17.8mm, APPROX.
Catalog No. 50-194-5328 Supplier Bellco Glass, Inc. Supplier No. 202900018
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 18-415, DIMENSIONS: OD OF CAP: 20.5mm, APPROX, ID OF CAP: 16.1mm, APROX, LENGTH: 17.8mm, APPROX.