Learn More
Bellco Glass, Inc. Blk Phenolic Screw Cap, 24mm Long Skirted - With Liner
SDP
Catalog No. 501945329
Encompass_Preferred
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 24-415, DIMENSIONS: OD OF CAP: 27.1mm, APPROX, ID OF CAP: 22.1mm, APROX, LENGTH: 18.7mm, APPROX.
Catalog No. 50-194-5329 Supplier Bellco Glass, Inc. Supplier No. 202900025
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 24-415, DIMENSIONS: OD OF CAP: 27.1mm, APPROX, ID OF CAP: 22.1mm, APROX, LENGTH: 18.7mm, APPROX.