Learn More
Bellco Glass, Inc. Blk Phenolic Screw Cap, 28mm Long Skirted - With Liner
SDP
Catalog No. 501945330
Encompass_Preferred
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 28-415, DIMENSIONS: OD OF CAP: 31.1mm, APPROX, ID OF CAP: 25.5mm, APROX, LENGTH: 20.3mm, APPROX.
Catalog No. 50-194-5330 Supplier Bellco Glass, Inc. Supplier No. 202900028
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 28-415, DIMENSIONS: OD OF CAP: 31.1mm, APPROX, ID OF CAP: 25.5mm, APROX, LENGTH: 20.3mm, APPROX.