Learn More
Bellco Glass, Inc. Blk Phenolic Screw Cap,38mm Long Skirted - with Liner
SDP
Catalog No. 501945331
Encompass_Preferred
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 38-415, DIMENSIONS: OD: 41.5mm, approx, ID: 38mm, approx, Length: 37.6mm, approx.
Catalog No. 50-194-5331 Supplier Bellco Glass, Inc. Supplier No. 202900038
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. The deep skirt reduces risk of contamination. 14B WHITE RUBBER (FDA CLASSIFICATION), THREAD FINISH:GPI 38-415, DIMENSIONS: OD: 41.5mm, approx, ID: 38mm, approx, Length: 37.6mm, approx.