Learn More
Bellco Glass, Inc. Blk Phenolic Screw Cap, 38mm With white rubber liner
SDP
Catalog No. 501945748
Encompass_Preferred
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and stem autoclaving. They have a non-toxic white rubber liner for all general culture applications. Packed 72 per case. MADE IN THE USA
Catalog No. 50-194-5748 Supplier Bellco Glass, Inc. Supplier No. 563638430
Black phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and stem autoclaving. They have a non-toxic white rubber liner for all general culture applications. Packed 72 per case. MADE IN THE USA