Learn More
Bellco Glass, Inc. Blk S/Cap W/Red PTFE/Sil,13mm With Septa
SDP
Catalog No. 501945383
Encompass_Preferred
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 13mm
Catalog No. 50-194-5383 Supplier Bellco Glass, Inc. Supplier No. 207000800
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 13mm