Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP7351EC10UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His-SUMO. Host: E. coli. Species Reactivity: Epstein-Barr. Protein Expression: A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Envelope glycoprotein GP350, was expressed in the hosts and tags indicated. AA Sequence: See company website. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 80% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: BLLF1. Gene ID: 3783713. Uniprot: P03200.
Catalog No. 50-188-7014