Learn More
Enzo Life Sciences Blm10 (Saccharomyces cerevisiae) polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLPW05700025
Encompass_Preferred
Immunogen: Recombinant N-terminal protein fragment (aa 1-157) of Saccharomyces cervisiae Blm10. Species reactivity: Saccharomyces cerevisiae. Applications: WB. Formulation: Liquid. Neat serum diluted in PBS containing 0.01% sodium azide.
Catalog No. 50-201-1539