Learn More
Sigma Aldrich Fine Chemicals Biosciences Bln I Avr II from Brevibac
Catalog No. 502905102
Encompass
Bln I recognizes the sequence C8595CTAGG and generates fragments with 5-cohesive termini.Compatible endsBln I ends are compatible with ends generated by Nhe I Spe I and Xba I.IsoschizomersBln I is an isoschizomer of Avr II.Note The complete 13 site Avr II restriction map of the E.coli genome has been reported.Methylation sensitivityThe enzyme is not known to be affected by methylation.
Catalog No. 50-290-5102 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11558170001
Bln I recognizes the sequence C8595CTAGG and generates fragments with 5-cohesive termini.Compatible endsBln I ends are compatible with ends generated by Nhe I Spe I and Xba I.IsoschizomersBln I is an isoschizomer of Avr II.Note The complete 13 site Avr II restriction map of the E.coli genome has been reported.Methylation sensitivityThe enzyme is not known to be affected by methylation.