Learn More
Aviva Systems Biology Corp BLNK Antibody - middle region (ARP46185_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46185_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BLNK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BLNK. Alternate names: BASH, BLNK-s, Ly57, MGC111051, SLP-65, SLP65, bca, AGM4, LY57, BLNK-SApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59263