Learn More
Aviva Systems Biology Corp Bloc1s2 Antibody - N-terminal region (ARP55703_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55703_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bloc1s2. It was validated on Western Blot. . Alternate names: 2410089B13Rik, BLOS2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-067