Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BLOC1S2 protein, human | >95.0% | 10 UG
SDP

Catalog No. 5000294128
Encompass_Preferred

Recombinant human BLOC1S2 protein (sequence M1-R99) is supplied as a tag-free fragment expressed in E. coli and purified for in vitro research. The protein has an approximate molecular mass of 11.35 kDa and is provided with high purity for biochemical assays, antibody generation, and interaction studies.

  • Tag-free recombinant fragment M1-R99.
  • Purity greater than 95% as determined by Tris-Bis PAGE.
  • Molecular weight approximately 11.35 kDa.
  • Expressed in Escherichia coli and purified.
  • Stable at 4°C for 1 week or at -20°C for extended storage; freeze aliquots for long term.
  • Suitable for biochemical assays, antibody production, and protein-protein interaction studies.

Catalog No. 50-002-94128 Supplier Medchemexpress LLC Supplier No. HYP76171A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human BLOC1S2 protein (sequence M1-R99) is supplied as a tag-free fragment expressed in E. coli and purified for in vitro research. The protein has an approximate molecular mass of 11.35 kDa and is provided with high purity for biochemical assays, antibody generation, and interaction studies.

  • Tag-free recombinant fragment M1-R99.
  • Purity greater than 95% as determined by Tris-Bis PAGE.
  • Molecular weight approximately 11.35 kDa.
  • Expressed in Escherichia coli and purified.
  • Stable at 4°C for 1 week or at -20°C for extended storage; freeze aliquots for long term.
  • Suitable for biochemical assays, antibody production, and protein-protein interaction studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.