Learn More
Medchemexpress LLC BLOC1S2 protein (recombinant, N-terminal GST fusion) | 97.0% | 50 UG
SDP
Catalog No. 5000294152
Encompass_Preferred
Recombinant human BLOC1S2 protein (N-terminal GST fusion) expressed in Escherichia coli and supplied as a purified 50 μg preparation. The GST tag facilitates affinity purification and detection, and the product is intended for biochemical assays, antibody generation, and protein-protein interaction studies. Store frozen at -20 °C and aliquot for long-term storage.
  • Recombinant fragment corresponding to residues M1-R99.
  • N-terminal GST tag for affinity purification and detection.
  • High purity (~97.0%) suitable for analytical and preparative use.
  • Approximate molecular weight ~34 kDa (tag plus fragment).
  • Expressed in Escherichia coli and provided in convenient laboratory quantities.
  • Stable when stored at -20 °C; aliquot to minimize freeze-thaw cycles.
Catalog No. 50-002-94152 Supplier Medchemexpress LLC Supplier No. HYP76171A50UG
Recombinant human BLOC1S2 protein (N-terminal GST fusion) expressed in Escherichia coli and supplied as a purified 50 μg preparation. The GST tag facilitates affinity purification and detection, and the product is intended for biochemical assays, antibody generation, and protein-protein interaction studies. Store frozen at -20 °C and aliquot for long-term storage.
  • Recombinant fragment corresponding to residues M1-R99.
  • N-terminal GST tag for affinity purification and detection.
  • High purity (~97.0%) suitable for analytical and preparative use.
  • Approximate molecular weight ~34 kDa (tag plus fragment).
  • Expressed in Escherichia coli and provided in convenient laboratory quantities.
  • Stable when stored at -20 °C; aliquot to minimize freeze-thaw cycles.