Learn More
Thermo Scientific™ 96-well Half/Full-Skirted PCR Plate Block, EMD
SCI_ED
Catalog No. S37360 Shop All Thermo Scientific Products
Change view
Click to view available options
Dimensions (L x W x H):
76 x 124 x 55 mm
Catalog No. Dimensions (L x W x H)
S37360 76 x 124 x 55 mm
1 options
Catalog No. S37360 Supplier Thermo Scientific™ Supplier No. 88870121/EMD
Add to Cart
Add to Cart
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
TRUSTED_SUSTAINABILITY
Specifications
Description 96-well half / full-skirted PCR
For Use With (Equipment) Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds 1 x 96 well half or full-skirted PCR plate
Type Block
Dimensions (L x W x H) 76 x 124 x 55 mm