Learn More
Cole-Parmer Stuart Block Heater Insert, 12 mm dia x 33 mm deep
Supplier:  Cole-Parmer 3661046
Encompass_Preferred
All blocks are made of anodized aluminum (except 36610-43). Most blocks feature a thermometer hole to let you easily monitor block temperature. Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Flexi-blocks use autoclavable high-density ceramic beads and are available in two sizes—double or triple width. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.
Catalog No. 50-196-0964