Learn More
Thomas Scientific Block, 12 x 5.0ml centrifuge tubes (17mm diameter)
SDP
Catalog No. 501458631
Encompass
The Digital DryBaths offer digital control over both temperature and time, eliminating the need for an external thermometer or timer. Available in three configurations, the digital dry baths offer capacity for up to four blocks. The cavities within the high grade aluminum blocks are precision-machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50ml tube sizes, providing unsurpassed temperature uniformity. Nine assorted blocks are available in total for compatibility with nearly all common tube sizes. All models include a convenient block lifter, for transferring blocks with high temperatures.
Catalog No. 50-145-8631 Supplier Thomas Scientific Supplier No. 1159N80
The Digital DryBaths offer digital control over both temperature and time, eliminating the need for an external thermometer or timer. Available in three configurations, the digital dry baths offer capacity for up to four blocks. The cavities within the high grade aluminum blocks are precision-machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50ml tube sizes, providing unsurpassed temperature uniformity. Nine assorted blocks are available in total for compatibility with nearly all common tube sizes. All models include a convenient block lifter, for transferring blocks with high temperatures.