Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for 17 - 18 mm Test Tubes, 12 Wells, 2081Q
SDP
Catalog No. 5032287
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 12 Wells
Model: 2081Q
Opening: 0.75" (1.9 cm)
Catalog No. 50-322-87 Supplier Electron Microscopy Sciences Supplier No. 6312905
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 12 Wells
Model: 2081Q
Opening: 0.75" (1.9 cm)