Learn More
Thomas Scientific Block 24 X 1.5/2.0ml Centrifuge Tubes
SDP
Catalog No. 501460112
Encompass
The Digital DryBaths offer digital control over both temperature and time, eliminating the need for an external thermometer or timer. Available in three configurations, the digital dry baths offer capacity for up to four blocks. The cavities within the high grade aluminum blocks are precision-machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50ml tube sizes, providing unsurpassed temperature uniformity. Nine assorted blocks are available in total for compatibility with nearly all common tube sizes. All models include a convenient block lifter, for transferring blocks with high temperatures.
Catalog No. 50-146-0112 Supplier Thomas Scientific Supplier No. 1227U85
The Digital DryBaths offer digital control over both temperature and time, eliminating the need for an external thermometer or timer. Available in three configurations, the digital dry baths offer capacity for up to four blocks. The cavities within the high grade aluminum blocks are precision-machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50ml tube sizes, providing unsurpassed temperature uniformity. Nine assorted blocks are available in total for compatibility with nearly all common tube sizes. All models include a convenient block lifter, for transferring blocks with high temperatures.