Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block Combination (3) 25mm; (5) 12-13mm; (6) 6mm, 14 Wells, 2076Q
SDP
Catalog No. 5032275
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 14 Wells
Model: 2076Q
Catalog No. 50-322-75 Supplier Electron Microscopy Sciences Supplier No. 6312910
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 14 Wells
Model: 2076Q