Learn More
Grainger Block 35 X 1.5Ml
Catalog No. 502751654
Encompass_Preferred
The Benchmark Scientific H5000-15 is a horizontal laminar flow clean bench that provides a particle-free work environment. It has a stainless steel interior with rounded corners for easy cleaning. The airflow is HEPA filtered and moves horizontally across the work surface at 90 feet per minute to sweep particles away. It has an LED lighting system and a removable perforated stainless steel work surface. The overall dimensions are 48"W x 24"D x 35.5"H. This clean bench is ETL certified to meet safety standards in the US and Canada.
Catalog No. 50-275-1654 Supplier Grainger Supplier No. 14N141
The Benchmark Scientific H5000-15 is a horizontal laminar flow clean bench that provides a particle-free work environment. It has a stainless steel interior with rounded corners for easy cleaning. The airflow is HEPA filtered and moves horizontally across the work surface at 90 feet per minute to sweep particles away. It has an LED lighting system and a removable perforated stainless steel work surface. The overall dimensions are 48"W x 24"D x 35.5"H. This clean bench is ETL certified to meet safety standards in the US and Canada.