Learn More
Grainger Block 35 X 2.0Ml
Catalog No. 502752703
Encompass_Preferred
The Grainger Approved Block 35 X 2.0Ml is a block of 35 2.0ml microcentrifuge tubes made of polypropylene. The tubes have attached domed caps and are graduated. They are sterile, DNase/RNase free, non-pyrogenic and non-cytotoxic. The tubes have a working volume of 1.5ml and can withstand centrifugal forces up to 20000 x g. The tubes have a shelf life of 5 years when stored at room temperature. The tubes are sold in packs of 500.
Catalog No. 50-275-2703 Supplier Grainger Supplier No. 14N142
The Grainger Approved Block 35 X 2.0Ml is a block of 35 2.0ml microcentrifuge tubes made of polypropylene. The tubes have attached domed caps and are graduated. They are sterile, DNase/RNase free, non-pyrogenic and non-cytotoxic. The tubes have a working volume of 1.5ml and can withstand centrifugal forces up to 20000 x g. The tubes have a shelf life of 5 years when stored at room temperature. The tubes are sold in packs of 500.