Learn More
Thomas Scientific Block, 48 X 0.2 ml PCR Tubes or 6 X 0.2 ml Strips
SDP
Catalog No. 501460165
Encompass
A broad temperature range, to 150ÝC, makes the Digital Dry Baths useful for a variety of applications in molecular biology, histology, clinical, environmental and industrial laboratories. A microprocessor regulates the high wattage heaters in the baths to provide precise, accurate control. The desired temperature is easily set using the arrow keys on the control panel while values are shown on the digital display. No checking of a thermometer and readjusting temperature is necessary. The user calibration feature allows for calibration to in house standards when required. Additionally, there is an RS232 port for recording block temperature activity. The block chamber is constructed of stainless steel, acting as a heat sink and providing a uniform transfer of heat from the heating elements to the interchangeable blocks. The resulting block uniformity ensures all samples receive the same temperature treatment, regardless of their block position. The AccuRack tube rack has capacity for up
Catalog No. 50-146-0165 Supplier Thomas Scientific Supplier No. 1228L05
A broad temperature range, to 150ÝC, makes the Digital Dry Baths useful for a variety of applications in molecular biology, histology, clinical, environmental and industrial laboratories. A microprocessor regulates the high wattage heaters in the baths to provide precise, accurate control. The desired temperature is easily set using the arrow keys on the control panel while values are shown on the digital display. No checking of a thermometer and readjusting temperature is necessary. The user calibration feature allows for calibration to in house standards when required. Additionally, there is an RS232 port for recording block temperature activity. The block chamber is constructed of stainless steel, acting as a heat sink and providing a uniform transfer of heat from the heating elements to the interchangeable blocks. The resulting block uniformity ensures all samples receive the same temperature treatment, regardless of their block position. The AccuRack tube rack has capacity for up