Learn More
Andwin Scientific BLOCK 50ML CENT TUBE
SDP
Catalog No. NC9653093
Encompass_Preferred
Modular Heating Block, 5-hole, holds Conical Bottom Centrifuge Tubes up to 50mL and 47.63mm dia. Solid aluminum, anodized blue modular block for producing maximum heat retention in and Signature Dry Block Heaters.
Catalog No. NC9653093 Supplier Andwin Scientific Supplier No. 13259254
Modular Heating Block, 5-hole, holds Conical Bottom Centrifuge Tubes up to 50mL and 47.63mm dia. Solid aluminum, anodized blue modular block for producing maximum heat retention in and Signature Dry Block Heaters.